HTL Labmate Pro Pipettors
Designed to make clinically demanding applications feel physically effortless
  • Contoured handle with finger-hook ensures correct grip, reduces fatigue and repetitive strain injury
  • Soft spring system enables smooth and easy pipetting with precision
  • Adjustable ejector height system accommodates nearly all brands of tips
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
Featuring a new pipetting mechanism, the intelligently crafted LABMATE PRO provides a true sense of comfort for laboratory professionals. UV-resistant and durable, these user-friendly pipettors offer outstanding accuracy and precision thanks to a dual volume setting using the pushbutton or the thumbwheel.
£197.30 - £568.14GBP / Each
£236.76 - £681.77GBP (incl VAT)
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (µL) Min Volume (µL)
Availability Pricing
HTL Labmate Pro LMP250 Single Channel Pipettor, Green; 50 to 250 µL 2455193 Mfr # 5660
COMPARE
Green 250 50
£197.30GBP / Each
£236.76GBP (incl VAT)
HTL Labmate Pro LMP2 Single Channel Pipettor, Red; 0.2 to 2 µL 2455194 Mfr # 5661
COMPARE
Red 2 0.2
£216.58GBP / Each
£259.90GBP (incl VAT)
HTL Labmate Pro LMP10 Single Channel Pipettor, Red; 0.5 to 10 µL 2455195 Mfr # 5662
COMPARE
Red 10 0.5
£216.58GBP / Each
£259.90GBP (incl VAT)
HTL Labmate Pro LMP20 Single Channel Pipettor, Yellow; 2 to 20 µL 2455196 Mfr # 5663
COMPARE
Yellow 20 2
£197.30GBP / Each
£236.76GBP (incl VAT)
HTL Labmate Pro LMP100 Single Channel Pipettor, Yellow; 10 to 100 µL 2455197 Mfr # 5664
COMPARE
Yellow 100 10
£197.30GBP / Each
£236.76GBP (incl VAT)
HTL Labmate Pro LMP200 Single Channel Pipettor, Yellow; 20 to 200 µL 2455198 Mfr # 5665
COMPARE
Yellow 200 20
£197.30GBP / Each
£236.76GBP (incl VAT)
HTL Labmate Pro LMP1000 Single Channel Pipettor, Blue; 100 to 1000 µL 2455199 Mfr # 5666
COMPARE
Blue 1000 100
£197.30GBP / Each
£236.76GBP (incl VAT)
HTL Labmate Pro LMP50 Single Channel Pipettor, Yellow; 5 to 50 µL 2455200 Mfr # 5667
COMPARE
Yellow 50 5
£197.30GBP / Each
£236.76GBP (incl VAT)
HTL Labmate Pro LMP5000 Single Channel Pipettor, White; 500 to 5000 µL 2455201 Mfr # 5668
COMPARE
White 5000 500
£232.32GBP / Each
£278.78GBP (incl VAT)
HTL Labmate Pro LMP10000 Single Channel Pipettor, White; 1000 to 10000 µL 2455202 Mfr # 5669
COMPARE
White 10000 1000
£246.87GBP / Each
£296.24GBP (incl VAT)
HTL Labmate Pro LMP8-10 Multichannel Pipettor, Red, 8-Channel; 0.5 to 10 µL 2455209 Mfr # 6281
COMPARE
Red 10 0.5
£418.31GBP / Each
£501.97GBP (incl VAT)
HTL Labmate Pro LMP8-50 Multichannel Pipettor, Yellow, 8-Channel; 5 to 50 µL 2455210 Mfr # 6282
COMPARE
Yellow 50 5
£418.31GBP / Each
£501.97GBP (incl VAT)
HTL Labmate Pro LMP8-200 Multichannel Pipettor, Yellow, 8-Channel; 20 to 200 µL 2455211 Mfr # 6283
COMPARE
Yellow 200 20
£418.31GBP / Each
£501.97GBP (incl VAT)
HTL Labmate Pro LMP8-300 Multichannel Pipettor, Green, 8-Channel; 50 to 300 µL 2455212 Mfr # 6284
COMPARE
Green 300 50
£418.31GBP / Each
£501.97GBP (incl VAT)
HTL Labmate Pro LMP12-10 Multichannel Pipettor, Red, 12-Channel; 0.5 to 10 µL 2455213 Mfr # 6285
COMPARE
Red 10 0.5
£568.14GBP / Each
£681.77GBP (incl VAT)
HTL Labmate Pro LMP12-50 Multichannel Pipettor, Yellow, 12-Channel; 5 to 50 µL 2455214 Mfr # 6286
COMPARE
Yellow 50 5
£568.14GBP / Each
£681.77GBP (incl VAT)
HTL Labmate Pro LMP12-200 Multichannel Pipettor, Yellow, 12-Channel; 20 to 200 µL 2455215 Mfr # 6287
COMPARE
Yellow 200 20
£568.14GBP / Each
£681.77GBP (incl VAT)
HTL Labmate Pro LMP12-300 Multichannel Pipettor, Green, 12-Channel; 50 to 300 µL 2455216 Mfr # 6288
COMPARE
Green 300 50
£568.14GBP / Each
£681.77GBP (incl VAT)
Showing 1- 18 of 18