HTL Labmate Pro LMP2 Single Channel Pipettor, Red; 0.2 to 2 µL
Representative image only.
HTL Labmate Pro LMP2 Single Channel Pipettor, Red; 0.2 to 2 µL
| Mfr: 5661| Item: 2455194
Designed to make clinically demanding applications feel physically effortless
£216.58GBP / Each
£259.90GBP (incl VAT)
£216.58GBP / Each
£259.90GBP (incl VAT)
Product Description
Key Features
  • Contoured handle with finger-hook ensures correct grip, reduces fatigue and repetitive strain injury
  • Soft spring system enables smooth and easy pipetting with precision
  • Adjustable ejector height system accommodates nearly all brands of tips
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
  • 3 year warranty
More About this Item
Featuring a new pipetting mechanism, the intelligently crafted LABMATE PRO provides a true sense of comfort for laboratory professionals. UV-resistant and durable, these user-friendly pipettors offer outstanding accuracy and precision thanks to a dual volume setting using the pushbutton or the thumbwheel.
3 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)2
  • Min Volume (µL)0.2
  • Accuracy (%)1.5
  • Precision (%)0.7
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeRed
  • DescriptionLabmate Pro LMP2 Single Channel Pipettor, Red; 0.2 to 2 µL