HTL Labmate Pro LMP8-200 Multichannel Pipettor, Yellow, 8-Channel; 20 to 200 µL
Representative image only.
HTL Labmate Pro LMP8-200 Multichannel Pipettor, Yellow, 8-Channel; 20 to 200 µL
| Mfr: 6283| Item: 2455211
Designed to make clinically demanding applications feel physically effortless
£418.31GBP / Each
£501.97GBP (incl VAT)
£418.31GBP / Each
£501.97GBP (incl VAT)
Product Description
Key Features
  • Contoured handle with finger-hook ensures correct grip, reduces fatigue and repetitive strain injury
  • Soft spring system enables smooth and easy pipetting with precision
  • Adjustable ejector height system accommodates nearly all brands of tips
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
  • 3 year warranty
More About this Item
Featuring a new pipetting mechanism, the intelligently crafted LABMATE PRO provides a true sense of comfort for laboratory professionals. UV-resistant and durable, these user-friendly pipettors offer outstanding accuracy and precision thanks to a dual volume setting using the pushbutton or the thumbwheel.
3 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)200
  • Min Volume (µL)20
  • Accuracy (%)1
  • Precision (%)0.6
  • Number Of Channels8
  • Tip EjectorYes
  • Color CodeYellow
  • DescriptionLabmate Pro LMP8-200 Multichannel Pipettor, Yellow, 8-Channel; 20 to 200 µL