HTL Labmate Pro LMP10000 Single Channel Pipettor, White; 1000 to 10000 µL
Representative image only.
HTL Labmate Pro LMP10000 Single Channel Pipettor, White; 1000 to 10000 µL
| Mfr: 5669| Item: 2455202
Designed to make clinically demanding applications feel physically effortless
£246.87GBP / Each
£296.24GBP (incl VAT)
£246.87GBP / Each
£296.24GBP (incl VAT)
Product Description
Key Features
  • Contoured handle with finger-hook ensures correct grip, reduces fatigue and repetitive strain injury
  • Soft spring system enables smooth and easy pipetting with precision
  • Adjustable ejector height system accommodates nearly all brands of tips
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
  • 3 year warranty
More About this Item
Featuring a new pipetting mechanism, the intelligently crafted LABMATE PRO provides a true sense of comfort for laboratory professionals. UV-resistant and durable, these user-friendly pipettors offer outstanding accuracy and precision thanks to a dual volume setting using the pushbutton or the thumbwheel.
3 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)10000
  • Min Volume (µL)1000
  • Accuracy (%)0.5
  • Precision (%)0.2
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeWhite
  • DescriptionLabmate Pro LMP10000 Single Channel Pipettor, White; 1000 to 10000 µL