HTL Labmate Pro LMP12-300 Multichannel Pipettor, Green, 12-Channel; 50 to 300 µL
Representative image only.
HTL Labmate Pro LMP12-300 Multichannel Pipettor, Green, 12-Channel; 50 to 300 µL
| Mfr: 6288| Item: 2455216
Designed to make clinically demanding applications feel physically effortless
£568.14GBP / Each
£681.77GBP (incl VAT)
£568.14GBP / Each
£681.77GBP (incl VAT)
Product Description
Key Features
  • Contoured handle with finger-hook ensures correct grip, reduces fatigue and repetitive strain injury
  • Soft spring system enables smooth and easy pipetting with precision
  • Adjustable ejector height system accommodates nearly all brands of tips
  • Fully autoclavable
  • Single-, 8- and 12-channel versions available
  • 3 year warranty
More About this Item
Featuring a new pipetting mechanism, the intelligently crafted LABMATE PRO provides a true sense of comfort for laboratory professionals. UV-resistant and durable, these user-friendly pipettors offer outstanding accuracy and precision thanks to a dual volume setting using the pushbutton or the thumbwheel.
3 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)300
  • Min Volume (µL)50
  • Accuracy (%)1
  • Precision (%)0.6
  • Number Of Channels12
  • Tip EjectorYes
  • Color CodeGreen
  • DescriptionLabmate Pro LMP12-300 Multichannel Pipettor, Green, 12-Channel; 50 to 300 µL