Trajan SGE eVol® XR Digital Analytical Syringes
Digitally controlled electronic drive and an analytical syringe—all in one!
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow—up to 98 steps!
  • Use for both GC and LC injections
Create a more efficient workflow using these programmable digital syringes. Easily change XCHANGE® syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination. Calibration and password protection ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
Call For Price
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Needle Included Syringe Capacity (µL) Description
Availability Pricing
Trajan SGE eVol® XR Digital Analytical Syringe, 5 µL, Removable Needle, 25G, Bevel Tip 1794001 Mfr # 2910020
COMPARE
Yes 5 Digital Analytical Syringe, 5 µL, Removable Needle, 25G, Bevel Tip
Call For Price
Trajan SGE EVOL Electronic Syringe, 5 µL; No Needle Included 1794002 Mfr # 2910021
COMPARE
No 5 Electronic Syringe, 5 µL; No Needle Included
Call For Price
Trajan SGE EVOL Electronic Syringe, 50 µL, Removable Needle, 23G; Bevel Tip 1794003 Mfr # 2910022
COMPARE
Yes 50 Electronic Syringe, 50 µL, Removable Needle, 23G; Bevel Tip
Call For Price
Trajan SGE EVOL Electronic Syringe, 50 µL; No Needle Included 1794004 Mfr # 2910023
COMPARE
No 50 Electronic Syringe, 50 µL; No Needle Included
Call For Price
Trajan SGE eVol® XR Digital Analytical Syringe, 500 µL, Removable Needle, 23G, Bevel Tip 1794005 Mfr # 2910024
COMPARE
Yes 500 Digital Analytical Syringe, 500 µL, Removable Needle, 23G, Bevel Tip
Call For Price
Trajan SGE eVol® XR Digital Analytical Syringe, 500 µL, No Needle 1794006 Mfr # 2910025
COMPARE
No 500 Digital Analytical Syringe, 500 µL, No Needle
Call For Price
Trajan SGE eVol® XR Digital Analytical Syringe, 100 µL, Removable Needle, 25G, Bevel Tip 1794010 Mfr # 2910029
COMPARE
Yes 100 Digital Analytical Syringe, 100 µL, Removable Needle, 25G, Bevel Tip
Call For Price
Trajan SGE EVOL Electronic Syringe, 1 mL; No Needle Included 1794011 Mfr # 2910035
COMPARE
No 1000 Electronic Syringe, 1 mL; No Needle Included
Call For Price
Trajan SGE EVOL Electronic Syringe, 100 µL; No Needle Included 1794012 Mfr # 2910037
COMPARE
No 100 Electronic Syringe, 100 µL; No Needle Included
Call For Price
Trajan SGE EVOL Electronic Syringe, 5 µL, Removable Needle, 23G; Bevel Tip 1794013 Mfr # 2910320
COMPARE
Yes 5 Electronic Syringe, 5 µL, Removable Needle, 23G; Bevel Tip
Call For Price
Trajan SGE eVol® XR Digital Analytical Syringe, 50 µL, Removable Needle, 25G, Bevel Tip; 3/Pk 1794014 Mfr # 2910322
COMPARE
Yes 50 Digital Analytical Syringe, 50 µL, Removable Needle, 25G, Bevel Tip; 3/Pk
Call For Price
Trajan SGE EVOL Replacement Syringe, 500 µL, 23G, Bevel Tip; 3/pk 1794015 Mfr # 2910324
COMPARE
Yes 500 Replacement Syringe, 500 µL, 23G, Bevel Tip; 3/pk
Call For Price
Trajan SGE EVOL XR Replacement Syringe, 1 mL, 23G, Bevel Tip; 3/pk 1794017 Mfr # 2910335
COMPARE
Yes 1000 XR Replacement Syringe, 1 mL, 23G, Bevel Tip; 3/pk
Call For Price
Showing 1- 13 of 13