Trajan SGE eVol® XR Digital Analytical Syringe, 500 µL, No Needle
Representative image only.
Trajan SGE eVol® XR Digital Analytical Syringe, 500 µL, No Needle
Trajan | Mfr: 2910025| Item: 1794006
Digitally controlled electronic drive and an analytical syringe—all in one!
Call For Price
Call For Price
Buy It Together
Current Item
Trajan SGE eVol® XR Digital Analytical Syringe, 500 µL, No Needle
Trajan SGE eVol® XR Digital Analytical Syringe, 500 µL, No Needle
1794006
Call For Price
+
Trajan SGE EVOL Electronic Syringe Stand
1794000
Call For Price
=
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow—up to 98 steps!
  • Use for both GC and LC injections
  • Electronic syringe features a 500 µL capacity
  • Needle sold separately
More About this Item
Create a more efficient workflow using these programmable digital syringes. Easily change XCHANGE® syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination. Calibration and password protection ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
Available for sale in the Middle East, Europe, and United States only.
Specifications
  • Syringe TypeDigital
  • Syringe Capacity (µL)500
  • Needle IncludedNo
  • SterileNo
  • DescriptionDigital Analytical Syringe, 500 µL, No Needle