Trajan SGE EVOL Electronic Syringe, 50 µL, Removable Needle, 23G; Bevel Tip
Representative image only.
Trajan SGE EVOL Electronic Syringe, 50 µL, Removable Needle, 23G; Bevel Tip
Trajan | Mfr: 2910022| Item: 1794003
Digitally controlled electronic drive and an analytical syringe—all in one!
Call For Price
Call For Price
Buy It Together
Current Item
Trajan SGE EVOL Electronic Syringe, 50 µL, Removable Needle, 23G; Bevel Tip
Trajan SGE EVOL Electronic Syringe, 50 µL, Removable Needle, 23G; Bevel Tip
1794003
Call For Price
+
Trajan SGE EVOL Electronic Syringe Stand
1794000
Call For Price
=
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow—up to 98 steps!
  • Use for both GC and LC injections
  • Volume: 50 µL Needle: 23G, Bevel Tip Pack size: 1
More About this Item
Create a more efficient workflow using these programmable digital syringes. Easily change XCHANGE® syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination. Calibration and password protection ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
Create a more efficient workflow using these programmable digital syringes. Easily exchange syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination. Calibration feature and password protection feature ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
Available for sale in the Middle East, Europe, and United States only.
Specifications
  • Syringe Capacity (µL)50
  • Needle IncludedYes
  • Needle Tip StyleBevel
  • Needle Gauge23
  • SterileNo
  • DescriptionElectronic Syringe, 50 µL, Removable Needle, 23G; Bevel Tip