Trajan SGE EVOL Electronic Syringe, 1 mL; No Needle Included
Representative image only.
Trajan SGE EVOL Electronic Syringe, 1 mL; No Needle Included
Trajan | Mfr: 2910035| Item: 1794011
Digitally controlled electronic drive and an analytical syringe—all in one!
Call For Price
Call For Price
Buy It Together
Current Item
Trajan SGE EVOL Electronic Syringe, 1 mL; No Needle Included
Trajan SGE EVOL Electronic Syringe, 1 mL; No Needle Included
1794011
Call For Price
+
Trajan SGE EVOL Electronic Syringe Stand
1794000
Call For Price
=
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow—up to 98 steps!
  • Use for both GC and LC injections
  • Volume: 1 mL Needle: None Pack size: 1
More About this Item
Create a more efficient workflow using these programmable digital syringes. Easily change XCHANGE® syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination. Calibration and password protection ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
Create a more efficient workflow using these programmable digital syringes. Easily exchange syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination. Calibration feature and password protection feature ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
Available for sale in the Middle East, Europe, and United States only.
Specifications
  • Syringe Capacity (µL)1000
  • Needle IncludedNo
  • SterileNo
  • DescriptionElectronic Syringe, 1 mL; No Needle Included