NSI Lab Solutions Environmental Wastewater Inorganics PT Express™

On-demand proficiency test (PT) for when you need results fast
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
Call For Price

32 variations of this product are available.

See All Product Options Below
32 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Form Description
Availability Pricing
COMPARE
pH Liquid pH Wastewater PT Express™; 2/PK
Call For Price
COMPARE
TOX Liquid TOX Wastewater PT Express™; 2/PK
Call For Price
COMPARE
MBAs Liquid MBAs Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Color Liquid Color Wastewater PT Express™; 2/PK
Call For Price
COMPARE
BOD, CBOD, COD, TOC Liquid Demand Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Silica Liquid Silica Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Acidity Liquid Acidity Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Bromide Liquid Bromide Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Mercury Liquid Mercury Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Nitrite Liquid Nitrite Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Total Suspended Solids, Total Solids Liquid Residue Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Calcium, Calcium Hardness, Magnesium, Total hardness Liquid Hardness Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Alkalinity, Chloride, Conductivity, Fluoride, Potassium, Sodium, Sulfate, TDS Liquid Minerals Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Ionic Salts Liquid Salinity Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Turbidity Liquid Turbidity Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Fats, Oils, Grease Liquid Fats, Oils, and Grease (FOGs) by IR Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Perchlorate Liquid Perchlorate Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Total Petroleum Hydrocarbons Liquid SGT - HEM (TPH) Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Ignitability Liquid Ignitability Wastewater PT Express™; 2/PK
Call For Price
COMPARE
Total Residual Chlorine Liquid Total Residual Chlorine Wastewater PT Express™; 2/PK
Call For Price
Showing 1 - 20 of 32

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat