NSI Lab Solutions QCI-087B Mercury Wastewater PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCI-087B Mercury Wastewater PT Express™; 2/PK

NSI Lab Solutions | Item: 6120267

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • Supplied in duplicate for convenience and value
  • Two 21-mL concentrate set for determination of Mercury
  • Each ampule produces 2 L of sample
  • Formulated in the NELAC range of 3.0 to 30 µg/L

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Specifications
  • Element/IonMercury
  • FormLiquid
  • Volume (mL)21
  • Packaging2/PK
  • Storage SymbolStore between 15 and 30 °C
  • UN Number2031
  • Shelf Life (months)24
  • DescriptionMercury Wastewater PT Express™; 2/PK
Documents & Certificates
Reviews
Q&A