Pipette Service Quote Request
Please complete the Quote Request form below and a member of the Kinesis pipette service team will call you back. Alternatively, if you would like to discuss your pipette service and calibration requirements with a Kinesis representative you can do so by calling +44 (0)1480 277802.
* All fields are required unless indicated otherwise.
enter your general contact information
Service required
Indicate how many pipettes for each calibration type?
  • SINGLE CHANNEL
  • MultiChannel
UKAS calibration
KPS3-10-UKAS 3 volume, 10 reading gravimetric test at minimum, middle and maximum volume of the pipettes working range
KPS3-5-UKAS 3 volume, 5 reading gravimetric test at minimum, middle and maximum volume of the pipettes working range
KPSALLM-UKAS Multichannel UKAS calibration of all channels
KPS2-10-UKAS 2 volume, 10 reading gravimetric test at minimum, middle and maximum volume of the pipettes working range
KPS2-5-UKAS 2 volume, 5 reading gravimetric test at minimum, middle and maximum volume of the pipettes working range
General pipette calibration
KPS2-2 Single channel (2 volumes, 2 readings) – accuracy and linearity check
KPS2-4 Single channel (2 volumes, 4 readings) – accuracy, linearity and precision check
KPSALLM Multichannel calibration of all channels simultaneously at all calibration levels