Reagents® Sulfuric Acid 50% (v/v), Standardized

Manufactured using the highest quality raw materials in ISO 9001 facility
  • 1 + 1
  • Aqueous
  • Colorless liquid
  • Density: 1.49
  • Specific gravity: 1.479 to 1.499 at 20 °C (68 °F)
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: <0.1
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Sulfuric Acid, 50% (v/v), Standardized, Cubitainer® Package, 10 L 6228821 Mfr # C7306200-10F
COMPARE
Standardized Cubitainer® package 10000
Call For Price
Reagents Sulfuric Acid, 50% (v/v), Standardized, Clear Glass Bottle, 2.5 L 6228822 Mfr # C7306200-2.5D1
COMPARE
Standardized Clear glass bottle 2500
Call For Price
Reagents Sulfuric Acid, 50% (v/v), Standardized, Natural Poly Bottle, 500 mL 6228823 Mfr # C7306200-500A
COMPARE
Standardized Poly bottle 500
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat