Reagents Sulfuric Acid, 50% (v/v), Standardized, Clear Glass Bottle, 2.5 L

Representative image only.

Reagents Sulfuric Acid, 50% (v/v), Standardized, Clear Glass Bottle, 2.5 L

Reagents | Mfr: C7306200-2.5D1| Item: 6228822

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • 1 + 1
  • Aqueous
  • Colorless liquid
  • Density: 1.49
  • Specific gravity: 1.479 to 1.499 at 20 °C (68 °F)
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: <0.1
  • 2.5 L clear glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration50% v/v
  • ClassStandardized
  • FormLiquid
  • Volume (mL)2500
  • PackagingClear glass bottle
  • DOT InformationSULFURIC ACID SOLUTION, 8, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1830
  • Shelf Life (days)760
  • Shelf Life (months)60
  • DescriptionSulfuric Acid, 50% (v/v), Standardized, Clear Glass Bottle, 2.5 L
Documents & Certificates
Reviews
Q&A