Reagents Sulfuric Acid, 50% (v/v), Standardized, Cubitainer® Package, 10 L

Representative image only.

Reagents Sulfuric Acid, 50% (v/v), Standardized, Cubitainer® Package, 10 L

Reagents | Mfr: C7306200-10F| Item: 6228821

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • 1 + 1
  • Aqueous
  • Colorless liquid
  • Density: 1.49
  • Specific gravity: 1.479 to 1.499 at 20 °C (68 °F)
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: <0.1
  • 10 L Cubitainer® package

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration50% v/v
  • ClassStandardized
  • FormLiquid
  • Volume (mL)10000
  • PackagingCubitainer® package
  • DOT InformationSULFURIC ACID SOLUTION, 8, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1830
  • Shelf Life (days)760
  • Shelf Life (months)60
  • DescriptionSulfuric Acid, 50% (v/v), Standardized, Cubitainer® Package, 10 L
Documents & Certificates
Reviews
Q&A