Reagents® Sulfamic Acid, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • White to off-white solid
  • Density: 2.15
  • Boiling point: decomposes
  • Melting point: 205 °C (401 °F) with decomposition
  • Solubility information: 213 g/L at 20 °C (68 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Sulfamic Acid, ACS Reagent, Wide-Mouth Container, 100 g 6228352 Mfr # C2114300-100A6
COMPARE
Salts Wide-mouth container 100
Call For Price
Reagents Sulfamic Acid, ACS Reagent, Poly Pail, 10 kg 6228353 Mfr # C2114300-10KG
COMPARE
Salts Poly pail 10000
Call For Price
Reagents Sulfamic Acid, ACS Reagent, Natural Poly Container, 2.5 kg 6228354 Mfr # C2114300-2.5A5
COMPARE
Salts Poly container 2500
Call For Price
Reagents Sulfamic Acid, ACS Reagent, Drum, 50 lb 6228355 Mfr # C2114300-50F5
COMPARE
Salts Drum-reagent, drum 22680
Call For Price
Reagents Sulfamic Acid, ACS Reagent, Natural Poly Container, 6 lb 6228356 Mfr # C2114300-6LB
COMPARE
Salts Poly container 2722
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat