Reagents Sulfamic Acid, ACS Reagent, Wide-Mouth Container, 100 g

Representative image only.

Reagents Sulfamic Acid, ACS Reagent, Wide-Mouth Container, 100 g

Reagents | Mfr: C2114300-100A6| Item: 6228352

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White to off-white solid
  • Density: 2.15
  • Boiling point: decomposes
  • Melting point: 205 °C (401 °F) with decomposition
  • Solubility information: 213 g/L at 20 °C (68 °F)
  • 100 g wide-mouth container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS
  • Linear FormulaNH2SO3H
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)100
  • Formula Weight97.09
  • PackagingWide-mouth container
  • DOT InformationSULFAMIC ACID, 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number5329-14-6
  • UN NumberUN2967
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSulfamic Acid, ACS Reagent, Wide-Mouth Container, 100 g
Documents & Certificates
Reviews
Q&A