Reagents Sulfamic Acid, ACS Reagent, Poly Pail, 10 kg

Representative image only.

Reagents Sulfamic Acid, ACS Reagent, Poly Pail, 10 kg

Reagents | Mfr: C2114300-10KG| Item: 6228353

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White to off-white solid
  • Density: 2.15
  • Boiling point: decomposes
  • Melting point: 205 °C (401 °F) with decomposition
  • Solubility information: 213 g/L at 20 °C (68 °F)
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS
  • Linear FormulaNH2SO3H
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)10000
  • Formula Weight97.09
  • PackagingPoly pail
  • DOT InformationSULFAMIC ACID, 8, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN2967
  • CAS Number5329-14-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSulfamic Acid, ACS Reagent, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A