NSI Lab Solutions Environmental Microbiology Drinking Water PT Express™ Cultures
On-demand proficiency test (PT) for when you need results fast
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.
These environmental microbiology drinking water proficiency tests are designed for quantitative and/or qualitative identification of target microorganisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.
Environmental Microbiology Drinking Water PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Qualitative/Quantitative Presence/Absence Drinking Water PT Express™; 10/PK
Call For Price
COMPARE
Qualitative Enterococci Drinking Water PT Express™; 10/PK
Call For Price
COMPARE
Quantitative Enterococci Drinking Water PT Express™
Call For Price
COMPARE
Quantitative Coliform Drinking Water PT Express™
Call For Price
COMPARE
Quantitative Standard Plate Count Drinking Water PT Express™