NSI Lab Solutions MIC-QC14B Quantitative Enterococci Drinking Water PT Express™

Representative image only.

NSI Lab Solutions MIC-QC14B Quantitative Enterococci Drinking Water PT Express™

NSI Lab Solutions | Item: 0500155

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Includes

Two bottles of sterile hydration fluid.

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • For quantitative determination of Enterococci in the range of 20 to 1000 CFU/MPN per 100 mL
  • Designed for use with all Most Probable Number (MPN) and Membrane Filtration (MF) procedures
  • Sample derived from E. faecalis (NCTC 775)
  • Supplied in duplicate for convenience and value

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental microbiology drinking water proficiency tests are designed for quantitative and/or qualitative identification of target microorganisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.

Environmental Microbiology Drinking Water PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Specifications
  • OrganismEnterococci
  • Organism typeBacteria
  • Packaging2/PK
  • Product TypeDrinking Water PT Express
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid: store at room temperature
  • DescriptionQuantitative Enterococci Drinking Water PT Express™
Reviews
Q&A