NSI Lab Solutions MIC-QC4B Qualitative/Quantitative Presence/Absence Drinking Water PT Express™; 10/PK

Representative image only.

NSI Lab Solutions MIC-QC4B Qualitative/Quantitative Presence/Absence Drinking Water PT Express™; 10/PK

NSI Lab Solutions | Item: 0500153

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Includes

10 bottles of sterile hydration fluid.

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • Report qualitative absence/presence and quantitative results
  • All samples cultured in the range of 20 to 200 CFU
  • Strains include E. coli (NCTC 9001), E. aerogenes (NCTC 10006), and P. aeruginosa (NCTC 12951)

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental microbiology drinking water proficiency tests are designed for quantitative and/or qualitative identification of target microorganisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.

Environmental Microbiology Drinking Water PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Ten sample set for determination of total/fecal coliforms and E. coli in drinking water. The samples are designed to be compatible with all promulgated methods including Membrane Filtration (MF), Multiple Tube Fermentation (MTF), IDEXX Quanti-Tray®, Colilert®, and Colisure®. Derived from primary cultures, each set contains three samples of total coliform positive/fecal and E. coli negative, three samples of total coliform/E. coli/fecal coliform positive, three samples for coliform negative, and two blanks.

Specifications
  • OrganismColiform, E. coli
  • Organism typeBacteria
  • Packaging10/PK
  • Product TypeDrinking Water PT Express
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid: store at room temperature
  • DescriptionQualitative/Quantitative Presence/Absence Drinking Water PT Express™; 10/PK
Reviews
Q&A