Value Plastics Male Luer Integral Lock Ring to Plug Adapters
Seal luer ports with a secure and leak-proof connection
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Ideal for capping off fluid transfer system
  • Prevent leaks with integral lock ring mechanism
These Value Plastics Male Luer Integral Lock Ring to Plug Adapters are designed to easily plug female luer ports. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The integral lock ring is a feature built into the male luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. These adapters are available in various materials to accommodate various applications and compliance requirements.
Choose from plugs closed at the tip and plugs closed at the grip. Plugs closed at luer tip are designed to seal at the tip of the plug, where it connects with the female luer fitting, ideal when it’s crucial to prevent any leakage or contamination at the tip of the fitting. Plugs closed at the grip feature a sealing mechanism located closer to the body of the plug, where it connects with the mating female luer fitting. This design may be preferred in applications where the tip of the fitting needs to remain open or where the focus is on sealing at the base rather than the tip.
£11.03 - £17.59GBP / Pkg of 25
£13.24 - £21.11GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Luer Connection Type (Port 2) Regulatory Approvals
Availability Pricing
Value Plastics Luer Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Integral Lock x Ring Plug, Closed at Grip; 25/Pk 5012009 Mfr # LP4-9
COMPARE
Polycarbonate Plug, Closed at Grip RoHS
£13.92GBP / Pkg of 25
£16.70GBP (incl VAT)
Value Plastics Luer Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x Plug, Closed at Luer Tip; 25/Pk 5012074 Mfr # MTLLP-1
COMPARE
Nylon Plug, Closed at Luer Tip RoHS
£11.03GBP / Pkg of 25
£13.24GBP (incl VAT)
Value Plastics Luer Fitting, Straight Adapter, Transparent Polypropylene, Male Luer Integral Lock Ring x Plug, Closed at Luer Tip; 25/Pk 5012075 Mfr # MTLLP-6005
COMPARE
Polypropylene Plug, Closed at Luer Tip FDA
£17.59GBP / Pkg of 25
£21.11GBP (incl VAT)