Trajan SGE BPX 70 Phase Capillary Columns
Offer enhanced endurance without loss of sensitivity
  • SilGuard™ integrated guard column technology provides additional protection by reducing the amount of nonvolatiles entering the analytical column
  • Provides the best possible combination of performance, robustness, reproducibility, low bleed, and inertness
  • Available in a wide variety of phases, IDs, lengths, and film thicknesses
The SGE BPX line of GC capillary columns provides the best possible combination of performance, robustness, reproducibility, low bleed, and inertness. It offers a comprehensive range of columns for almost any chromatographic application, including chemical, environmental, food, and fuels.
£728.24 - £1,868.26GBP / Each
£873.89 - £2,241.91GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Film Thickness (µm) Phase
Availability Pricing
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.22 mm x 0.25 µm; 25 m Length 1794164 Mfr # 054602
COMPARE
0.22 0.25 BPX70
£744.41GBP / Each
£893.29GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.22 mm x 0.25 µm; 50 m Length 1794165 Mfr # 054603
COMPARE
0.22 0.25 BPX70
£1,282.67GBP / Each
£1,539.20GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.32 mm x 0.25 µm; 25 m length 1794166 Mfr # 54606
COMPARE
0.32 0.25 BPX70
£793.44GBP / Each
£952.13GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.32 mm x 0.25 µm; 50 m length 1794167 Mfr # 54607
COMPARE
0.32 0.25 BPX70
£1,340.55GBP / Each
£1,608.66GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.22 mm x 0.25 µm; 30 m Length 1794168 Mfr # 054612
COMPARE
0.22 0.25 BPX70
£746.38GBP / Each
£895.66GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.22 mm x 0.25 µm; 60 m Length 1794169 Mfr # 054613
COMPARE
0.22 0.25 BPX70
£1,301.40GBP / Each
£1,561.68GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.32 mm x 0.25 µm; 30 m length 1794170 Mfr # 54616
COMPARE
0.32 0.25 BPX70
£786.03GBP / Each
£943.24GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.32 mm x 0.25 µm; 60 m length 1794171 Mfr # 54617
COMPARE
0.32 0.25 BPX70
£1,359.27GBP / Each
£1,631.12GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 30 m length 1794173 Mfr # 54622
COMPARE
0.25 0.25 BPX70
£728.24GBP / Each
£873.89GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 60 m length 1794174 Mfr # 54623
COMPARE
0.25 0.25 BPX70
£1,263.09GBP / Each
£1,515.71GBP (incl VAT)
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 120 m length 1794175 Mfr # 54624
COMPARE
0.25 0.25 BPX70
£1,868.26GBP / Each
£2,241.91GBP (incl VAT)
Showing 1- 11 of 11