Spex Volatile Organics Mix with 60 Certified Components
Analytical standards for drinking water, wastewater, and solid waste
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 502.2 and 8260
US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Description
Availability Pricing
COMPARE
200 µg/mL (200 ppm) Volatile Organics Mix with 60 Certified Components, 200 µg/mL in Methanol-P&T; 1 mL
Call For Price
COMPARE
1,000 µg/mL (1,000 ppm) Volatile Organics Mix for US EPA Methods 502.2 and 8260 (60 Components), 1,000 µg/mL in Methanol-P&T; 1 mL
Call For Price
COMPARE
2,000 µg/mL (2,000 ppm) Volatile Organics Mix with 60 Certified Components, 2,000 µg/mL in Methanol-P&T; 1 mL