Spex 60-BIG-MIX-200 Volatile Organics Mix (60 Components), 200 µg/mL; 1 mL

Representative image only.

Spex 60-BIG-MIX-200 Volatile Organics Mix (60 Components), 200 µg/mL; 1 mL

Spex | Item: 0455689

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 502.2 and 8260
  • 200 ppm volatile organics mix in methanol-P&T matrix
  • 1 mL ampule

More About this Item

US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).

US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 60 components.

• Volatile Organics Mix is a 1 mL ampule which contains 200 µg/mL (200 ppm) of Benzene, Bromobenzene, Bromochloromethane, Bromodichloromethane, Bromoform, Bromomethane, n-Butylbenzene, sec-Butylbenzene, tert-Butylbenzene, Carbon tetrachloride, Chlorobenzene, Chloroethane, Chloroform, Chloromethane, 2-Chlorotoluene, 4-Chlorotoluene, 1,2-Dibromo-3-chloropropane, Dibromochloromethane, 1,2-Dibromoethane, Dibromomethane, 1,2-Dichlorobenzene, 1,3-Dichlorobenzene, 1,4-Dichlorobenzene, Dichlorodifluoromethane, 1,1-Dichloroethane, 1,2-Dichloroethane, 1,1-Dichloroethene, cis-1,2-Dichloroethene, trans-1,2-Dichloroethene, 1,2-Dichloropropane, 1,1-Dichloropropene, cis-1,3-Dichloropropene, trans-1,3-Dichloropropene, 1,3-Dichloropropane, 2,2-Dichloropropane, Ethylbenzene, Hexachlorobutadiene, Isopropylbenzene, p-Isopropyltoluene, Methylene chloride, Naphthalene, n-Propylbenzene, Styrene, 1,1,1,2-Tetrachloroethane, 1,1,2,2-Tetrachloroethane, Tetrachloroethene, Toluene, 1,2,3-Trichlorobenzene, 1,2,4-Trichlorobenzene, 1,1,1-Trichloroethane, 1,1,2-Trichloroethane, Trichloroethene, Trichlorofluoromethane, 1,2,3-Trichloropropane, 1,2,4-Trimethylbenzene, 1,3,5-Trimethylbenzene, Vinyl chloride, m-Xylene, o-Xylene, and p-Xylene.

Hazardous material shipping fees may apply.

Specifications
  • Concentration200 µg/mL (200 ppm)
  • MatrixMethanol-P&T
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number71-43-2, 108-86-1, 74-97-5, 75-27-4, 75-25-2, 74-83-9, 104-51-8, 135-98-8, 98-06-6, 56-23-5, 108-90-7, 75-00-3, 67-66-3, 74-87-3, 95-49-8, 106-43-4, 96-12-8, 124-48-1, 106-93-4, 74-95-3, 95-50-1, 541-73-1, 106-46-7, 75-71-8, 75-34-3, 107-06-2, 75-35-4, 156-59-2, 156-60-5, 78-87-5, 563-58-6, 10061-01-5, 10061-02-6, 142-28-9, 594-20-7, 100-41-4, 87-68-3, 98-82-8, 99-87-6, 75-09-2, 91-20-3, 103-65-1, 100-42-5, 630-20-6, 79-34-5, 127-18-4, 108-88-3, 87-61-6, 120-82-1, 71-55-6, 79-00-5, 79-01-6, 75-69-4, 96-18-4, 95-63-6, 108-67-8, 75-01-4, 108-38-3, 95-47-6, 106-42-3
  • UN NumberUN1230
  • Shelf Life (months)12
  • DescriptionVolatile Organics Mix with 60 Certified Components, 200 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A