Spex USP <561> Standards, 100 µg/mL
Tests and specifications for the safety of botanical products
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
The United States Pharmacopeia (USP) creates standards for the monitoring of pharmaceutical and botanical products. General Chapter <561> Articles of Botanical Origin, outlines tests and specifications for the safety of botanical products. The chapter outlines sampling procedures and tests for general characteristics, such as for moisture, ash, starch and fiber. The chapter also describes methods for extractables and volatile compounds. In the interests of safety, USP <561> has procedures for the testing of mycotoxins and designates targeted pesticides. SPEX CertiPrep provides standards for testing of mycotoxins and pesticides under the General Chapter <561>.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Matrix Description
Availability Pricing
COMPARE
alpha-BHC, beta-BHC, delta-BHC, Endosulfan sulfate, Endrin, Etrimfos, gamma-BHC, Heptachlor, Heptachlor epoxide (Isomer A), Heptachlor epoxide (Isomer B), Hexachlorobenzene, Malaoxon, Malathion, Methamidophos, Methidathion, Methoxychlor, Mirex, Monocrotophos, Parathion, tau-Fluvalinate, Vinclozolin Acetonitrile USP <561> Mix B, 100 µg/mL in Acetonitrile; 1 mL
Call For Price
COMPARE
1,2,4,5-Tetrachloro-3-nitrobenzene; 2,3,4,5,6-Pentachloroanisole; Bis(2,3,3,3-tetrachloropropyl) ether (S421); Dimethyl-p-Nitrophenylphosphate; Imidan (Phosmet); Methyl parathion; N-desethyl-primiphos-methyl; Paraoxon; Pentachloroaniline; Pentachloronitrobenzene; Permethrin; Phosalone; Piperonyl butoxide; Pirimiphos-ethyl; Pirimiphos-methyl; Procymidone; Profenofos; Prowl (Pendimethalin); Pyrethrins (mix of isomers); Quinalphos; Tetradifon; Tokuthion/Prothiophos Acetonitrile USP <561> Mix A, 100 µg/mL in Acetonitrile; 1 mL
Call For Price
COMPARE
Acarol (Bromopropylate), Azinphos-ethyl, Azinphos-Methyl, Baythroid (Cyfluthrin), Bromophos-ethyl, Bromophos-methyl, Chlorfenvinfos, Chlorpyrifos-ethyl, Chlorpyrifos-methyl, Cypermethrin, Dichlofluanid, Dyfonate (Fonofos), Ethion, Fenchlorphos, Fenitrothion, Fensulfothion, Fenthion, Kelthane (Dicofol), lambda-Cyhalothrin, Sanmarton Acetonitrile USP <561> Mix C, 100 µg/mL in Acetonitrile; 1 mL
Call For Price
COMPARE
Acephate; Alachlor; Aldrin; alpha-Chlordane; Deltamethrin; Diazinon; Dichlorvos; Dieldrin; Dimethoate; Endosulfan I; Endosulfan II; gamma-Chlordane; o,p'-DDD; o,p'-DDE; o,p'-DDT; Omethoate; Oxychlordane; p,p'-DDD; p,p'-DDE; p,p'-DDT Acetonitrile USP <561> Mix D, 100 µg/mL in Acetonitrile; 1 mL
Call For Price
COMPARE
Ziram Acetonitrile USP <561> Mix F, 100 µg/mL in Acetonitrile; 1 mL
Call For Price
COMPARE
Fenchlorophos-oxon, Fenpropathrin, Fensulfothion-oxon, Fensulfothion-oxon-sulfone, Fensulfothion-sulfone, Fenthion sulfoxide, Fenthion-oxon, Fenthion-oxon-sulfone, Fenthion-oxon-sulfoxide, Fenthion-sulfone, Flucythrinate, Mecarbam, Methacrifos, Methyl pentachlorophenyl sulfide LC/MS Acetonitrile USP <561> Mix E, 100 µg/mL in LC/MS Acetonitrile; 1 mL
Call For Price
COMPARE
Epsilon-HCH Toluene USP <561> Mix G, 100 µg/mL in Toluene; 1 mL
Call For Price
Showing 1- 7 of 7