Spex USP-561-C USP <561> Mix C, 100 µg/mL in Acetonitrile; 1 mL

Representative image only.

Spex USP-561-C USP <561> Mix C, 100 µg/mL in Acetonitrile; 1 mL

Spex | Item: 4105977

Tests and specifications for the safety of botanical products

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • 100 ppm USP <561> Mix C in acetonitrile matrix
  • 1 mL ampule

More About this Item

The United States Pharmacopeia (USP) creates standards for the monitoring of pharmaceutical and botanical products. General Chapter <561> Articles of Botanical Origin, outlines tests and specifications for the safety of botanical products. The chapter outlines sampling procedures and tests for general characteristics, such as for moisture, ash, starch and fiber. The chapter also describes methods for extractables and volatile compounds. In the interests of safety, USP <561> has procedures for the testing of mycotoxins and designates targeted pesticides. SPEX CertiPrep provides standards for testing of mycotoxins and pesticides under the General Chapter <561>.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration100 µg/mL (100 ppm)
  • MatrixAcetonitrile
  • Element/IonAcarol (Bromopropylate), Azinphos-ethyl, Azinphos-Methyl, Baythroid (Cyfluthrin), Bromophos-ethyl, Bromophos-methyl, Chlorfenvinfos, Chlorpyrifos-ethyl, Chlorpyrifos-methyl, Cypermethrin, Dichlofluanid, Dyfonate (Fonofos), Ethion, Fenchlorphos, Fenitrothion, Fensulfothion, Fenthion, Kelthane (Dicofol), lambda-Cyhalothrin, Sanmarton
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Acetonitrile; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Irritant
  • UN NumberUN1648
  • CAS Number18181-80-1, 2642-71-9, 86-50-0, 68359-37-5, 4824-78-6, 2104-96-3, 470-90-6, 39475-55-3, 5598-13-0, 52315-07-8, 1085-98-9, 944-22-9, 563-12-2, 299-84-3, 122-14-5, 115-90-2, 55-38-9, 115-32-2, 91465-08-6, 51630-58-1
  • Shelf Life (months)12
  • DescriptionUSP <561> Mix C, 100 µg/mL in Acetonitrile; 1 mL
Reviews
Q&A