Spex Sulfur Oil Standards for Diesel Fuel Analysis in Base Oil 20
For use in diesel fuel analysis
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Concentration Description
Availability Pricing
COMPARE
Blank Sulfur Oil Blank for Diesel Fuel Analysis, 0 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
5 µg/g (5 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 5 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
10 µg/g (10 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 10 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
15 µg/g (15 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 15 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
20 µg/g (20 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 20 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
25 µg/g (25 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 25 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
50 µg/g (50 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 50 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
75 µg/g (75 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 75 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
100 µg/g (100 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 100 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
200 µg/g (200 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 200 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
300 µg/g (300 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 300 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
500 µg/g (500 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 500 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
750 µg/g (750 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 750 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
1,000 µg/g (1,000 ppm) Sulfur Oil Standard for Diesel Fuel Analysis, 1,000 µg/g in Base Oil 20; 100 mL
Call For Price
COMPARE
Multiple Sulfur Oil Standard Set for Diesel Fuel Analysisin Base Oil 20, 8 Bottles
Call For Price
Showing 1- 15 of 15