Spex DSS8-SET Sulfur Oil Standard Set in Base Oil 20, 8 Bottles

Representative image only.

Spex DSS8-SET Sulfur Oil Standard Set in Base Oil 20, 8 Bottles

Spex | Item: 0457044

For use in diesel fuel analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Sulfur oil standard set in base oil 20 contains 8 standards

More About this Item

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This sulfur oil standard set contains 8 solutions.

• Solution 1 is a 100 mL bottle which contains 1,000 µg/g (1,000 ppm) of Sulfur. This is available individually as part number 04570-43.
• Solution 2 is a 100 mL bottle which contains 750 µg/g (750 ppm) of Sulfur. This is available individually as part number 04570-42.
• Solution 3 is a 100 mL bottle which contains 500 µg/g (500 ppm) of Sulfur. This is available individually as part number 04570-41.
• Solution 4 is a 100 mL bottle which contains 300 µg/g (300 ppm) of Sulfur. This is available individually as part number 04570-40.
• Solution 5 is a 100 mL bottle which contains 200 µg/g (200 ppm) of Sulfur. This is available individually as part number 04570-39.
• Solution 6 is a 100 mL bottle which contains 100 µg/g (100 ppm) of Sulfur. This is available individually as part number 04570-38.
• Solution 7 is a 100 mL bottle which contains 50 µg/g (50 ppm) of Sulfur. This is available individually as part number 04570-36.
• Solution 8 is a 500 mL bottle of base oil 20.

Specifications
  • ConcentrationMultiple
  • MatrixBase Oil 20
  • Element/IonEight bottle set
  • FormLiquid
  • Volume (mL)100
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionSulfur Oil Standard Set for Diesel Fuel Analysisin Base Oil 20, 8 Bottles
Documents & Certificates
Reviews
Q&A