Spex Multi-Element Organometallic Oil Standards (21 Elements)

Alleviate engine failures by analyzing metal suspended in the used oils
  • Guaranteed stable and accurate
  • Clear, transparent mix
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.

Applications include:
• Wear metals
• Crude oils
• Additive metals
• Environmental monitoring
• Petrochemical testing
• Pharmaceuticals
• Food processing
• Sulfur in diesel fuel

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Concentration Weight (g) Description
Availability Pricing
COMPARE
100 µg/g 50 Multi-Element Organometallic Oil Standard (21 Elements), 100 µg/g, in Base Oil 75; 50 g
Call For Price
COMPARE
100 µg/g 100 Multi-Element Organometallic Oil Standard (21 Elements), 100 µg/g, in Base Oil 75; 100 g
Call For Price
COMPARE
300 µg/g 50 Multi-Element Organometallic Oil Standard (21 Elements), 300 µg/g, in Base Oil 75; 50 g
Call For Price
COMPARE
300 µg/g 100 Multi-Element Organometallic Oil Standard (21 Elements), 300 µg/g, in Base Oil 75; 100 g
Call For Price
COMPARE
500 µg/g 50 Multi-Element Organometallic Oil Standard (21 Elements), 500 µg/g, in Base Oil 75; 50 g
Call For Price
COMPARE
500 µg/g 100 Multi-Element Organometallic Oil Standard (21 Elements), 500 µg/g, in Base Oil 75; 100 g
Call For Price
COMPARE
900 µg/g 50 Multi-Element Organometallic Oil Standard (21 Elements), 900 µg/g, in Base Oil 75; 50 g
Call For Price
COMPARE
900 µg/g 100 Multi-Element Organometallic Oil Standard (21 Elements), 900 µg/g, in Base Oil 75; 100 g
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat