Spex S21-300Z Multi-Element Organometallic Oil Standard (21 Elements), 300 µg/g, in Base Oil 75; 50 g

Representative image only.

Spex S21-300Z Multi-Element Organometallic Oil Standard (21 Elements), 300 µg/g, in Base Oil 75; 50 g

Spex | Item: 0456995

Alleviate engine failures by analyzing metal suspended in the used oils

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Guaranteed stable and accurate
  • Clear, transparent mix
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • 300 µg/g organometallic oil standard (21 elements) in base oil 75 matrix
  • 50 g bottle

More About this Item

The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.

Applications include:
• Wear metals
• Crude oils
• Additive metals
• Environmental monitoring
• Petrochemical testing
• Pharmaceuticals
• Food processing
• Sulfur in diesel fuel

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component standard contains 21 components.

• Multi-Element Organometallic Oil Standard (21 Elements) is a 50 g bottle which contains 300 µg/g of Aluminum, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, and Zinc.

Specifications
  • Concentration300 µg/g
  • MatrixBase Oil 75
  • Element/IonMultiple
  • FormLiquid
  • Weight (g)50
  • DOT InformationNon-Hazardous:: Not regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionMulti-Element Organometallic Oil Standard (21 Elements), 300 µg/g, in Base Oil 75; 50 g
Documents & Certificates
Reviews
Q&A