Spex Multi-Element Organometallic Oil Standard, 23 Elements
Multi-element standards for organometallic oil
  • Determine wear metals in engine oils and lubricants
  • ISO 9001 and ISO/IEC 17025 compliant
Spex CertiPrep® multi-element organometallic oil standards are designed to determine wear of metals in engine oils and other lubricants used in machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc.
By tracking metals suspended in the used oil, you can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.
Applications include wear metals, crude oils, additive metals, environmental monitoring, petrochemical testing, pharmaceuticals, food processing, and sulfur in diesel fuel.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Aluminum, Antimonhy, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Potassium, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, Zinc 50 Multi-Element Organometallic Oil Standards, 23 Elements, 100 µg/g, in Base Oil 75; 50 g
Call For Price
COMPARE
Aluminum, Antimonhy, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Potassium, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, Zinc 100000 Multi-Element Organometallic Oil Standards, 23 Elements, 100 µg/g, in Base Oil 75; 100 g
Call For Price
COMPARE
Aluminum, Antimonhy, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Potassium, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, Zinc 50 Multi-Element Organometallic Oil Standards, 23 Elements, 300 µg/g, in Base Oil 75; 50 g
Call For Price
COMPARE
Aluminum, Antimonhy, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Potassium, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, Zinc 100 Multi-Element Organometallic Oil Standards, 23 Elements, 300 µg/g, in Base Oil 75; 100 g
Call For Price
COMPARE
Aluminum, Antimonhy, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Potassium, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, Zinc 50 Multi-Element Organometallic Oil Standards, 23 Elements, 500 µg/g, in Base Oil 75; 50 g
Call For Price
COMPARE
Aluminum, Antimonhy, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Potassium, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, Zinc 100 Multi-Element Organometallic Oil Standards, 23 Elements, 500 µg/g, in Base Oil 75; 100 g
Call For Price
COMPARE
Aluminum, Antimonhy, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Potassium, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, Zinc 50 Multi-Element Organometallic Oil Standards, 23 Elements, 900 µg/g, in Base Oil 75; 50 g
Call For Price
COMPARE
Aluminum, Antimonhy, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Potassium, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, Zinc 100 Multi-Element Organometallic Oil Standards, 23 Elements, 900 µg/g, in Base Oil 75; 100 g
Call For Price
Showing 1- 8 of 8