Spex S23-500Z Multi-Element Organometallic Oil Standards, 23 Elements, 500 µg/g, in Base Oil 75; 50 g

Representative image only.

Spex S23-500Z Multi-Element Organometallic Oil Standards, 23 Elements, 500 µg/g, in Base Oil 75; 50 g

Spex | Item: 0456989

Multi-element standards for organometallic oil

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Determine wear metals in engine oils and lubricants
  • ISO 9001 and ISO/IEC 17025 compliant

More About this Item

Spex CertiPrep® multi-element organometallic oil standards are designed to determine wear of metals in engine oils and other lubricants used in machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc.

By tracking metals suspended in the used oil, you can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.

Applications include wear metals, crude oils, additive metals, environmental monitoring, petrochemical testing, pharmaceuticals, food processing, and sulfur in diesel fuel.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Specifications
  • Concentration500 µg/g
  • MatrixBase oil 75
  • Element/IonAluminum, Antimonhy, Barium, Boron, Cadmium, Calcium, Chromium, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Phosphorus, Potassium, Silicon, Silver, Sodium, Tin, Titanium, Vanadium, Zinc
  • FormLiquid
  • Volume (mL)50
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • UN NumberNot regulated
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionMulti-Element Organometallic Oil Standards, 23 Elements, 500 µg/g, in Base Oil 75; 50 g
Documents & Certificates
Reviews
Q&A