Spex EU 20813 Standards, 100 µg/mL
Pesticide standards for use with the EU monograph for monitoring pesticides in herbal drugs and preparations
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • European Pharmacopeia Monograph 2.8.13 designation
The European Pharmacopeia Monograph 2.8.13 designates targeted pesticides for monitoring in herbal drugs and herbal preparations. This list targets several dozen common pesticides in use around the world and limits their concentration in herbal products to the low ppm and ppb range. SPEX CertiPrep has created pesticide standards for use with the EU monograph to help the analyst detect pesticides at their critical action levels.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Matrix Description
Availability Pricing
COMPARE
Acephate; Alachlor; Aldrin; alpha-Chlordane; Deltamethrin; Diazinon; Dichlorvos; Dieldrin; Dimethoate; Endosulfan I; Endosulfan II; gamma-Chlordane; o,p'-DDD; o,p'-DDE; o,p'-DDT; Omethoate; Oxychlordane; p,p'-DDD; p,p'-DDE; p,p'-DDT Acetonitrile EU 20813 Mix E, 100 µg/mL in LC/MS Acetonitrile; 1 mL
Call For Price
COMPARE
1,2,4,5-Tetrachloro-3-nitrobenzene; 2,3,4,5,6-Pentachloroanisole; Bis(2,3,3,3-tetrachloropropyl) ether (S421); Dimethyl-p-Nitrophenylphosphate; Imidan (Phosmet); Methyl parathion; N-desethyl-primiphos-methyl; Paraoxon; Pentachloroaniline; Pentachloronitrobenzene; Permethrin; Phosalone; Piperonyl butoxide; Pirimiphos-ethyl; Pirimiphos-methyl; Procymidone; Profenofos; Prowl (Pendimethalin); Pyrethrins (mix of isomers); Quinalphos; Tetradifon; Tokuthion/Prothiophos Acetonitrile EU 20813 Mix A, 100 µg/mL in Acetonitrile; 1 mL
Call For Price
COMPARE
Epsilon-HCH Toluene EU 20813 Mix G, 100 µg/mL in Toluene; 1 mL
Call For Price
COMPARE
Ziram Acetonitrile EU 20813 Mix D, 100 µg/mL in Acetonitrile; 1 mL
Call For Price
COMPARE
Acarol (Bromopropylate), Azinphos-ethyl, Azinphos-Methyl, Baythroid (Cyfluthrin), Bromophos-ethyl, Bromophos-methyl, Chlorfenvinfos, Chlorpyrifos-ethyl, Chlorpyrifos-methyl, Cypermethrin, Dichlofluanid, Dyfonate (Fonofos), Ethion, Fenchlorphos, Fenitrothion, Fensulfothion, Fenthion, Kelthane (Dicofol), lambda-Cyhalothrin, Sanmarton Acetonitrile EU 20813 Mix C, 100 µg/mL in Acetonitrile; 1 mL
Call For Price
COMPARE
alpha-BHC, beta-BHC, delta-BHC, Endosulfan sulfate, Endrin, Etrimfos, gamma-BHC, Heptachlor, Heptachlor epoxide (Isomer A), Heptachlor epoxide (Isomer B), Hexachlorobenzene, Malaoxon, Malathion, Methamidophos, Methidathion, Methoxychlor, Mirex, Monocrotophos, Parathion, tau-Fluvalinate, Vinclozolin Acetonitrile EU 20813 Mix B, 100 µg/mL in Acetonitrile; 1 mL