Spex EU 20813 Mix C, 100 µg/mL in Acetonitrile; 1 mL

Representative image only.

Spex EU 20813 Mix C, 100 µg/mL in Acetonitrile; 1 mL

Spex | Item: 4106018

Pesticide standards for use with the EU monograph for monitoring pesticides in herbal drugs and preparations

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • European Pharmacopeia Monograph 2.8.13 designation
  • 100 ppm EU 20813 Mix C in acetonitrile matrix
  • 1 mL ampule

More About this Item

The European Pharmacopeia Monograph 2.8.13 designates targeted pesticides for monitoring in herbal drugs and herbal preparations. This list targets several dozen common pesticides in use around the world and limits their concentration in herbal products to the low ppm and ppb range. SPEX CertiPrep has created pesticide standards for use with the EU monograph to help the analyst detect pesticides at their critical action levels.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration100 µg/mL (100 ppm)
  • MatrixAcetonitrile
  • Element/IonAcarol (Bromopropylate), Azinphos-ethyl, Azinphos-Methyl, Baythroid (Cyfluthrin), Bromophos-ethyl, Bromophos-methyl, Chlorfenvinfos, Chlorpyrifos-ethyl, Chlorpyrifos-methyl, Cypermethrin, Dichlofluanid, Dyfonate (Fonofos), Ethion, Fenchlorphos, Fenitrothion, Fensulfothion, Fenthion, Kelthane (Dicofol), lambda-Cyhalothrin, Sanmarton
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Acetonitrile; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Irritant
  • UN NumberUN1648
  • CAS Number18181-80-1, 2642-71-9, 86-50-0, 68359-37-5, 4824-78-6, 2104-96-3, 470-90-6, 39475-55-3, 5598-13-0, 52315-07-8, 1085-98-9, 944-22-9, 563-12-2, 299-84-3, 122-14-5, 115-90-2, 55-38-9, 115-32-2, 91465-08-6, 51630-58-1
  • Shelf Life (months)12
  • DescriptionEU 20813 Mix C, 100 µg/mL in Acetonitrile; 1 mL
Reviews
Q&A