Spex Claritas PPT® Grade ICP-MS Spike Sample Standards
Multi-element standards for ICP-MS analysis
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Designed for addition to a matrix blank prior to digestion for both water and soil
  • CLP-M/6020/SW-846 standards for ICP-MS
SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation.
Our spike standards, 04568-93 and 04568-94, provide all of the analytes required for ICP-MS. Add 1 mL of 04568-93 to aqueous samples and 2 mL of 04568-94 to solid samples prior to digestion.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
Multiple 125 Claritas PPT® Grade ICP-MS Spike Sample Standard 1 (Water) in 5% HNO3/tr. Tartaric Acid/tr. HF; 125mL
Call For Price
COMPARE
Multiple 125 Claritas PPT® Grade ICP-MS Spike Sample Standard 2 (Soil) in 5% HNO3/tr. Tartaric Acid/tr. HF; 125 mL