Spex CL-SPIKE-1 Claritas PPT® Grade ICP-MS Spike Sample Standard 1 (Water); 125mL

Representative image only.

Spex CL-SPIKE-1 Claritas PPT® Grade ICP-MS Spike Sample Standard 1 (Water); 125mL

Spex | Item: 0456893

Multi-element standards for ICP-MS analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Designed for addition to a matrix blank prior to digestion for both water and soil
  • CLP-M/6020/SW-846 standards for ICP-MS
  • Spike sample standard 1 in 5% HNO3/tr. Tartaric Acid/tr. HF matrix
  • 125 mL bottle

More About this Item

SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation.

Our spike standards, 04568-93 and 04568-94, provide all of the analytes required for ICP-MS. Add 1 mL of 04568-93 to aqueous samples and 2 mL of 04568-94 to solid samples prior to digestion.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 17 elements.

• Spike sample standard 1 is a 125 mL bottle of 500 µg/mL (500 ppm) Iron; 250 µg/mL (250 ppm) Barium, Zinc; 100 µg/mL (100 ppm) Antimony, Chromium, Cobalt, Copper, Manganese, Nickel, Vanadium; 50 µg/mL (50 ppm) Arsenic, Lead; and 25 µg/mL (25 ppm) Beryllium, Cadmium, Selenium, Silver, Thallium in 5% HNO3/tr. Tartaric Acid/tr. HF.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeClaritas PPT® Grade
  • ConcentrationMultiple
  • Matrix5% HNO3/tr. Tartaric Acid/tr. HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • Polarity Index9
  • Solubility in Water (% wt/wt)100
  • Specific Gravity1
  • UV Cutoff (nm)200
  • DescriptionClaritas PPT® Grade ICP-MS Spike Sample Standard 1 (Water) in 5% HNO3/tr. Tartaric Acid/tr. HF; 125mL
Documents & Certificates
Reviews
Q&A