Spex Claritas PPT® Grade ICP-MS Instrument Check Standards
Multi-element standards for ICP and ICP-MS instrumentation
  • Calibrate ICP-MS instrumentation prior to analysis
  • Dilute to the concentration appropriate for the instrument
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
Spex CertiPrep® Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.
Our Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS. Spex CertiPrep®Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
£156.00 - £389.00GBP / Each
£187.20 - £466.80GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Aluminum, Antimony, Arsenic, Barium, Beryllium, Cadmium, Chromium, Cobalt, Copper, Lead, Manganese, Nickel, Selenium, Silver, Thallium, Vanadium, Zinc 125 Claritas PPT® Grade ICP-MS Instrument Check Standard 1, 10 µg/mL in 2% HNO3/tr. Tartaric Acid/tr. HF; 125 mL
£389.00GBP / Each
£466.80GBP (incl VAT)
COMPARE
Molybdenum, Thorium, Uranium 125 Claritas PPT® Grade ICP-MS Instrument Check Standard 4, 10 µg/mL in 2% HNO3; 125 mL
Call For Price
COMPARE
Calcium, Iron, Magnesium, Potassium, Sodium, 125 Claritas PPT® Grade ICP-MS Instrument Check Standard 3, 200 µg/mL in 2% HNO3; 125 mL
Call For Price
COMPARE
Molybdenum, Strontium, Tin, Titanium, 125 Claritas PPT® Grade ICP-MS Instrument Check Standard 5, 10 µg/mL in 2% HNO3/tr. HF; 125 mL
£156.00GBP / Each
£187.20GBP (incl VAT)
COMPARE
Aluminum, Antimony, Arsenic, Barium, Beryllium, Cadmium, Chromium, Cobalt, Copper, Lead, Manganese, Nickel, Salenium, Silver, Thallium, Vandium, Zinc, Molybdenum, Thorium, Uranium, Calcium, Iron, Magnesium, Potassium, Sodium, Strontium, Tin, Titanium, Mercury 125 Claritas PPT® Grade ICP-MS Instrument Check Standards Set; 125 mL