Spex CL-ICS-4 Claritas PPT® Grade ICP-MS Instrument Check Standard 4, 10 µg/mL in 2% HNO3; 125 mL

Representative image only.

Spex CL-ICS-4 Claritas PPT® Grade ICP-MS Instrument Check Standard 4, 10 µg/mL in 2% HNO3; 125 mL

Spex | Item: 0456876

Multi-element standards for ICP and ICP-MS instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Calibrate ICP-MS instrumentation prior to analysis
  • Dilute to the concentration appropriate for the instrument
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • 10 ppm internal standard 4 in 2% HNO3 matrix

More About this Item

Spex CertiPrep® Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.

Our Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS. Spex CertiPrep®Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10 µg/mL (10 ppm)
  • Purity GradeClaritas PPT® Grade
  • Matrix2% HNO3
  • ClassICP-MS
  • Element/IonMolybdenum, Thorium, Uranium
  • FormLiquid
  • Volume (mL)125
  • DOT InformationCorrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8; Packaging Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionClaritas PPT® Grade ICP-MS Instrument Check Standard 4, 10 µg/mL in 2% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A