Spex Assurance® Grade TCLP Standards for AA & ICP
Multi-element standards for AA and ICP analysis
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2019
  • For use in accordance with the Toxicity Characteristic Rule Regulatory Levels issues in the Federal Register 55, 11846 March 1990; Method 1311
Designed to determine the mobility of the Inorganic contaminants present in liquid, solid and multi-phase wastes. To simplify, TCLP is designed to determine the hazardous contaminants that are actually entering into the environment. In addition to the SPEX CertiPrep TCLP TCLP Standards, designed with all of the elements in one solution, the Toxicity Characteristic rule separates the elements according to specific instrumentation: ICP, GFAA, and Cold Vapor AA.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Multiple 125 Assurance® Grade TCLP Standard with Mercury for AA & ICP in 2% HNO3; 125 mL
Call For Price
COMPARE
Multiple 500 Assurance® Grade TCLP Standard with Mercury for AA & ICP in 2% HNO3; 500 mL
Call For Price
COMPARE
Multiple 125 Assurance® Grade TCLP Standard without Mercury for AA & ICP in 2% HNO3; 125 mL
Call For Price
COMPARE
Multiple 500 Assurance® Grade TCLP Standard without Mercury for AA & ICP in 2% HNO3; 500 mL