Spex TCLP-100 Assurance® Grade TCLP Standard with Mercury for AA & ICP; 125 mL

Representative image only.

Spex TCLP-100 Assurance® Grade TCLP Standard with Mercury for AA & ICP; 125 mL

Spex | Item: 0456829

Multi-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2019
  • For use in accordance with the Toxicity Characteristic Rule Regulatory Levels issues in the Federal Register 55, 11846 March 1990; Method 1311
  • Toxicity Characteristic Leachate Performance (TCLP) standard with mercury for AA & ICP in 2% HNO3 matrix
  • 125 mL bottle

More About this Item

Designed to determine the mobility of the Inorganic contaminants present in liquid, solid and multi-phase wastes. To simplify, TCLP is designed to determine the hazardous contaminants that are actually entering into the environment. In addition to the SPEX CertiPrep TCLP TCLP Standards, designed with all of the elements in one solution, the Toxicity Characteristic rule separates the elements according to specific instrumentation: ICP, GFAA, and Cold Vapor AA.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 8 elements.

• TCLP Standard is a 125 mL bottle which contains 500 µg/mL (500 ppm) of Barium; 100 µg/mL (100 ppm) of Mercury*; 25 µg/mL (25 ppm) of Arsenic, Chromium, Lead, Silver; and 5 µg/mL (5 ppm) of Cadmium, Selenium.

Hazardous material shipping fees may apply.

*Mercury is supplied as a separate solution in 5% HNO3 due to incompatibility with other elements.

Specifications
  • Purity GradeAssurance® Grade
  • ConcentrationMultiple
  • Matrix2% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade TCLP Standard with Mercury for AA & ICP in 2% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A