Spex Assurance® Grade Gold, 1,000 µg/mL
Single-element standards for AA and ICP analysis
  • Made with acid and ASTM Type I water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. They are packaged in 30 mL, 125 mL, and 500 mL bottles to minimize contamination. Custom standards can be manufactured upon request.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Element/Ion Volume (mL)
Availability Pricing
COMPARE
1,000 µg/mL (1,000 ppm) Gold 125
Call For Price
COMPARE
1,000 µg/mL (1,000 ppm) Gold 30
Call For Price
COMPARE
1,000 µg/mL (1,000 ppm) Gold 500