Spex PLAU3-2M Assurance® Grade Gold, 1,000 µg/mL (1,000 ppm) for AA and ICP in 10% HCl, 30 mL

Representative image only.

Spex PLAU3-2M Assurance® Grade Gold, 1,000 µg/mL (1,000 ppm) for AA and ICP in 10% HCl, 30 mL

Spex | Item: 0450480

Single-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • 30 mL bottle

More About this Item

Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. They are packaged in 30 mL, 125 mL, and 500 mL bottles to minimize contamination. Custom standards can be manufactured upon request.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves.
If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Specific treatment (see on this label). Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.

Irritant
Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • Purity GradeAssurance® Grade
  • Empirical FormulaAu
  • Matrixin 10% HCl
  • ClassInorganic Standards
  • Element/IonGold
  • Volume (mL)30
  • DOT InformationNon-Hazardous
  • Hazard SymbolIrritant
  • UN NumberUN1789
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionGold, 1,000 µg/mL (1,000 ppm) for AA and ICP in 10% HCl, 30 mL
Reviews
Q&A