Spex Assurance® Grade Calibration Standard Sets
Multi-element standards for AA & ICP instrumentation
  • Calibrate AA and ICP instrumentation prior to analysis
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition)
AA & ICP Calibration Standards are provided for routine instrument calibration. The concentrations and matrices have been selected for convenience of use and stability.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
Multiple 125 Assurance® Grade Calibration Standard Set with Mercury for AA & ICP, 6 Bottle Set; 125 mL
Call For Price
COMPARE
Multiple 125 Assurance® Grade Calibration Standard Set without Mercury for AA & ICP, 5 Bottle Set; 125 mL
Call For Price
COMPARE
Multiple 125 Assurance® Grade Mixed Calibration Standard Set for AA & ICP, 5 Bottle Set; 125 mL