Spex MXSTD-SETAN Assurance® Grade Calib. Std Set w/o Mercury for AA & ICP; 125 mL

Representative image only.

Spex MXSTD-SETAN Assurance® Grade Calib. Std Set w/o Mercury for AA & ICP; 125 mL

Spex | Item: 0456774

Multi-element standards for AA & ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Calibrate AA and ICP instrumentation prior to analysis
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition)
  • Calibration standard without mercury for AA & ICP contains 5 solutions
  • Five 125 mL bottles

More About this Item

AA & ICP Calibration Standards are provided for routine instrument calibration. The concentrations and matrices have been selected for convenience of use and stability.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This mixed multi-element calibration standard contains 5 solutions.

• Solution 1 is a 125 mL bottle which contains 100 µg/mL (100 ppm) of Arsenic, Calcium; 50 µg/mL (50 ppm) of Antimony, Selenium; 20 µg/mL (20 ppm) of Boron, Cadmium, Copper; 10 µg/mL (10 ppm) of Barium; and 5 µg/mL (5 ppm) of Silver in 5% HNO3/tr. Tartaric Acid/tr. HF. This is available individually as part number 04567-59.

• Solution 2 is a 125 mL bottle which contains 200 µg/mL (200 ppm) of Potassium; 100 µg/mL (100 ppm) of Molybdenum, Sodium; 50 µg/mL (50 ppm) of Lithium; and 10 µg/mL (10 ppm) of Strontium in 5% HNO3. This is available individually as part number 04567-63.

• Solution 3 is a 125 mL bottle which contains 100 µg/mL (100 ppm) of Phosphorus and 20 µg/mL (20 ppm) of Cerium, Cobalt, Vanadium in 5% HNO3. This is available individually as part number 04567-65.

• Solution 4 is a 125 mL bottle which contains 100 µg/mL (100 ppm) of Aluminum, Silica, Titanium; 50 µg/mL (50 ppm) of Chromium, Zinc; and 40 µg/mL (40 ppm) of Tin in 5% HNO3/tr. HF. This is available individually as part number 04567-69.

• Solution 5 is a 125 mL bottle which contains 100 µg/mL (100 ppm) of Iron, Lead, Magnesium; 50 µg/mL (50 ppm) of Thallium; 20 µg/mL (20 ppm) of Nickel; and 10 µg/mL (10 ppm) of Beryllium in 5% HNO3. This is available individually as part number 04567-71.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeAssurance® Grade
  • ConcentrationMultiple
  • MatrixMultiple
  • Element/IonFive bottle set
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Calibration Standard Set without Mercury for AA & ICP, 5 Bottle Set; 125 mL
Documents & Certificates
Reviews
Q&A