Spex Assurance® Grade Calcium, 10,000 µg/mL

Single-element standards for AA and ICP analysis
  • Made with acid and ASTM Type I water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Custom standards can be manufactured upon request.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Note: May cause severe skin burns and serious eye damage. Do not breathe dusts or mists. If on skin or hair, rinse with water/shower and immediately take off all contaminated clothing. If in eyes, rinse cautiously with water for several minutes, remove contact lenses if present and easy to do so, and continue rinsing. Immediately call a poison center and doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Concentration Matrix Volume (mL)
Availability Pricing
COMPARE
10,000 µg/mL (10,000 ppm) 5% HNO3 125
Call For Price
COMPARE
10,000 µg/mL (10,000 ppm) 5% HNO3 250
Call For Price
COMPARE
10,000 µg/mL (10,000 ppm) 5% HNO3 500
Call For Price
COMPARE
10,000 µg/mL (10,000 ppm) 5% HCI 500
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat