Spex PLCA2-3Y Assurance® Grade Calcium, 10,000 µg/mL, for AA and ICP in 5% HNO3; 125 mL

Representative image only.

Spex PLCA2-3Y Assurance® Grade Calcium, 10,000 µg/mL, for AA and ICP in 5% HNO3; 125 mL

Spex | Item: 0450319

Single-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • 10,000 ppm Calcium in 5% HNO3 matrix

More About this Item

Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Custom standards can be manufactured upon request.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Note: May cause severe skin burns and serious eye damage. Do not breathe dusts or mists. If on skin or hair, rinse with water/shower and immediately take off all contaminated clothing. If in eyes, rinse cautiously with water for several minutes, remove contact lenses if present and easy to do so, and continue rinsing. Immediately call a poison center and doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10,000 µg/mL (10,000 ppm)
  • Matrix5% HNO3
  • Element/IonCalcium
  • FormLiquid
  • Volume (mL)125
  • DOT InformationCorrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8; Packaging Group III
  • Storage SymbolAmbient
  • UN NumberUN3264
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionSPEX CertiPrep Assurance® Grade Calcium, 10,000 µg/mL for AA and ICP in 5% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A