Spex Acrolein and Acrylonitrile Standards
Analytical standards for drinking water, wastewater, and solid waste
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 603
US EPA Method 603 is a general analytical method for the separation of acrolein and acrylonitrile in municipal and industrial wastewater by GC/FID.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Concentration Description
Availability Pricing
COMPARE
107-02-8, 107-13-1 2,000 µg/mL (2,000 ppm) Acrolein and Acrylonitrile Standard, 2,000 µg/mL in Methanol-P&T; 1 mL
Call For Price
COMPARE
107-02-8, 107-13-1 10,000 µg/mL (10,000 ppm) Acrolein and Acrylonitrile Standard, 10,000 µg/mL in Methanol-P&T; 1 mL