Spex 603-XM Acrolein and Acrylonitrile Standard, 2,000 µg/mL; 1 mL

Representative image only.

Spex 603-XM Acrolein and Acrylonitrile Standard, 2,000 µg/mL; 1 mL

Spex | Item: 0455652

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 603
  • 2,000 ppm Acrolein and Acrylonitrile standard in methanol-P&T matrix
  • 1 mL ampule

More About this Item

US EPA Method 603 is a general analytical method for the separation of acrolein and acrylonitrile in municipal and industrial wastewater by GC/FID.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 2 components.

• Acrolein and Acrylonitrile Standard is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of Acrolein and Acrylonitrile.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol-P&T
  • Element/IonAcrolein, Acrylonitrile
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Corrosion, Acute Toxicity, Irritant, Health Hazard
  • UN NumberUN1230
  • CAS Number107-02-8, 107-13-1
  • Shelf Life (months)2
  • DescriptionAcrolein and Acrylonitrile Standard, 2,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A