Reagents® Sulfuric Acid, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Assay 95 to 98%
  • Colorless liquid
  • Density: 1.84
  • Solubility information: miscible
  • pH range: <1
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Description
Availability Pricing
Reagents Sulfuric Acid, ACS Reagent, Amber Glass Bottle, 1 L 6228646 Mfr # C5102600-1C
COMPARE
Acid Amber glass bottle Sulfuric Acid, ACS Reagent, Amber Glass Bottle, 1 L
Call For Price
Reagents Sulfuric Acid, ACS Reagent, Winpak Container, 75 lb 6228647 Mfr # C5102600-75LB
COMPARE
Acid Winpak container Sulfuric Acid, ACS Reagent, Winpak Container, 75 lb
Call For Price
Reagents Sulfuric Acid, ACS Reagent, Natural Poly Bottle, 1 L 6228648 Mfr # C5102640-1A
COMPARE
Acid Poly bottle Sulfuric Acid, ACS Reagent, Natural Poly Bottle, 1 L
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat